Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Phire Plant Direct PCR Master Mix PROMO
Amplify DNA directly from plant samples without prior DNA purification with Phire Plant Direct PCR Master Mix.
Specifications
GC-Rich PCR Performance | Medium |
---|---|
PCR Method | Direct PCR |
Polymerase | Phire Hot Start II DNA polymerase |
Reaction Speed | Fast |
Product Line | Phire |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
F160SPM
|
Thermo Scientific™
F160S |
250 x 50 μL Reactions | N/A | N/A | N/A | ||||
Description
Highlights
- Direct PCR - sample is added directly to PCR reaction, therefore there is no need for time-consuming and expensive DNA purification steps.
- Dilution and Storage protocol available - allows tens of PCR reactions from one tiny sample and allows re-testing
- Specially engineered Phire Hot Start II DNA polymerase - extremely short PCR protocol times
- Master Mix format with premixed gel loading dye - minimizes possibility of cross-contamination, reduces sample handling and allows direct loading on gel
Applications
- Genotyping
- Transgene detection
- Knockout analysis
- Sequencing
Includes
- 2x Phire Plant Direct PCR Master Mix
- Dilution Buffer
- Control Primer Mix (25 μM each)
- Water, nuclease- free
- O'GeneRuler Express DNA Ladder
Specifications
Medium | |
Phire Hot Start II DNA polymerase | |
Phire | |
Genotyping | |
Blunt | |
Plant |
Direct PCR | |
Fast | |
Direct PCR Master Mix | |
Built-In Hot Start | |
SuperMix or Master Mix | |
DNA |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title