Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phire Tissue Direct PCR Master Mix PROMO

Amplify DNA directly from variety of human and animal samples with no prior purification with Phire Tissue Direct PCR Master Mix.

Supplier:  Thermo Scientific™ F170S

 View more versions of this product

Catalog No. F170SPM


Description

Description

The Phire Tissue Direct PCR Master Mix has been developed for amplification of DNA directly from a wide variety of tissues obtained from mice, human, fish, birds and insects. The Master Mix containing Phire Hot Start II DNA Polymerase is specially formulated to perform PCR in the presence of different animal tissue-derived inhibitors such as collagen, melanin and eumelanin (hair, skin) or myoglobin (muscle). The kit also includes Thermo Scientific DNARelease Additive, which can be used to improve the release of DNA from difficult tissues.

Highlights

  • Direct PCR – sample is added directly to PCR reaction, therefore therese is no need for time-consuming and expensive DNA purification steps
  • Dilution and Storage protocol available – allows tens of PCR reactions from one tiny sample and allows re-testing
  • Specially engineered Phire DNA polymerase – extremely short PCR protocol times
  • Master Mix format with premixed gel loading dye – minimizes possibility of cross-contamination, reduces sample handling and allows direct loading on gel

Applications

  • Genotyping
  • Transgene detection
  • Knockout analysis
  • Sequencing

Includes

  • 2X Phire Tissue Direct PCR Master Mix
  • Reaction voume with dilution and storage protocol: 20 μL
  • Reaction voume with direct protocol: 50 μL
  • Dilution Buffer
  • DNARelease Additive
  • Universal control primer mix (25 μM each)
  • Water, nuclease- free
  • O'GeneRuler Express DNA Ladder
Specifications

Specifications

Contains: 2 x 1.25 mL Phire Tissue Direct PCR Master Mix (2X), 5 mL Dilution Buffer, 3 x 100 μL DNARelease Additive, 40 μL Control Primer Mix (25 μM each), 2 x 1.25 mL Water, nuclease- free, 50 μg O'GeneRuler Express DNA Ladder (sufficient for 100 applications), Store at -20°C.
Direct PCR
Fast
Phire
250 x 20 μL Reactions
Built-In Hot Start
Blunt
Tissue
Medium
Phire Hot Start II DNA polymerase
Yes
Direct PCR Master Mix
Genotyping
250 x 20 mL Reactions
SuperMix or Master Mix
DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only. Not for use in diagnostic procedures.