Learn More
Description
Specifications
Specifications
| Antigen | PHLDA1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Alias | Apoptosis-associated nuclear protein, DT1P1B11, MGC131738, PHRIPT-cell death-associated gene 51 protein, pleckstrin homology-like domain family A member 1, pleckstrin homology-like domain, family A, member 1, PQ-rich protein, Proline- and glutamine-rich protein, Proline- and histidine-rich protein, proline-histidine rich protein, TDAG51PQR protein |
| Gene Symbols | PHLDA1 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
