Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHLPP Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB164988 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164988 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB164988 Supplier Novus Biologicals Supplier No. NBP281766JF549
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PHLPP Polyclonal antibody specifically detects PHLPP in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen PHLPP
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias EC 3.1.3.16, hSCOP, KIAA0606, MGC161555, PH domain and leucine rich repeat protein phosphatase, PH domain and leucine rich repeat protein phosphatase 1, PH domain leucine-rich repeat-containing protein phosphatase 1, PHLPP, pleckstrin homology domain containing, family E (with leucine rich repeats)member 1, Pleckstrin homology domain-containing family E member 1, PLEKHE1, SCN circadian oscillatory protein, SCOPPH domain-containing family E member 1, Suprachiasmatic nucleus circadian oscillatory protein
Host Species Rabbit
Immunogen PHLPP antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human PHLPP. The immunogen is located within amino acids 250 - 300 of PHLPP. Amino Acid Squence: REPAEPPPEAGPRLAPPE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Protein Phosphatase, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 23239
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.