Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Phorbol-12-myristate-13-acetate, >99%

Catalog No. p-7083168
Click to view available options
Quantity:
1 mg
10 mg
25 mg

Phorbol-12-myristate-13-acetate, >99% is a potent tumor promoter and activates protein kinase C in vivo and in vitro.

  • PMA′s effects on PKC are attributed to its similarity to diacylgylerol, a natural activator of PKC.
  • PMA activates Ca2+− ATPase and potentiates forskolin-induced cAMP formation.
  • It has been shown to inhibit apoptosis induced by the Fas antigen, but PMA induces apoptosis in HL-60 promyelocytic leukemia cells.
TRUSTED_SUSTAINABILITY

Specifications

Purity >98% (HPLC)
For Use With (Application) Potent tumor promoter, Activates protein kinase C in vivo and in vitro
Solubility Practically insoluble.
Molecular Formula C36H56O8
CAS 16561-29-8
Content And Storage Store at − 20°C. Protect from light.
Quantity 1 mg
Molecular Weight (g/mol) 616.84 g/mol
Melting Point 162°F
Vapor Pressure 1.8X10-17 mm Hg at 25°C
Form Solid
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.