Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phorbol 12-myristate 13-acetate, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
5 mg
Description
Phorbol 12-myristate 13-acetate is an extensively used phorbol ester high affinity activator of protein kinase C (Ki = 2.6 nM as measured by displacement of [3H]phorbol 12,13-dibutyrate binding in rat cortex synaptosomal membranes). It binds to the C1a and C1b domains of PKC inducing membrane translocation of PKC. Phorbol 12-myristate 13-acetate activates the PKC isoforms PKCα, -β, -γ, -δ, -ε, -η, and -θ but not PKCζ or -ι/λ. Phorbol 12-myristate 13-acetate is a tumor promoter. Phorbol 12-myristate 13-acetate can also induce differentiation of THP-1 monocytes to a macrophage phenotype in vitro.
Specifications
Quantity | 1 mg |
Formula Weight | 616.83 |
Percent Purity | >99% |
Chemical Name or Material | Phorbol 12-myristate 13-acetate |
Safety and Handling
Recommended Storage | Store at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction