Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Phortress

Click to view available options
Quantity:
10 mg
50 mg
Description
Phortress is a prodrug of the antitumor agent 5F 203, which acts via binding to aryl hydrocarbon receptors. Induces expression of CYP1A1 and generates adducts in the DNA of sensitive MCF7 and IGROV-1 cells.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | Phortress |
| Purity | >98% |
| Molecular Weight (g/mol) | 459.41 |
| Inhibitors | Phortress |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction