Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Phortress
GREENER_CHOICE

Catalog No. p-5545961
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Prodrug of the antitumor agent 5F 203

Phortress is a prodrug of the antitumor agent 5F 203, which acts via binding to aryl hydrocarbon receptors. Induces expression of CYP1A1 and generates adducts in the DNA of sensitive MCF7 and IGROV-1 cells.

Specifications

Quantity 10 mg
Product Type Phortress
Purity >98%
Molecular Weight (g/mol) 459.41
Inhibitors Phortress

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.