Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphate, 0.2 M Buffer Solution, pH 4.4, PROMO, Thermo Scientific Chemicals

Catalog No. J60467APPM
Click to view available options
Quantity:
500 mL
1 L

Widely used in molecular biology, biochemistry and chromatography. Useful in conjunction with Sodium Phosphate, dibasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.2 M
Molecular Weight (g/mol) 119.98 g/mol
Quantity 500 mL
Packaging Plastic Bottle
CAS 7558-80-7
Content And Storage Keep cold
Form Liquid

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.