Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphate Buffer, 0.05 Molar, pH 7.4, Reagents
Click to view available options
Quantity:
4 L
20 L
Packaging:
Cubitainer
HDPE Winpak
Poly Bottle
Specifications
Specifications
Color | Colorless |
pH | 7.4 |
Quantity | 20 L |
Packaging | HDPE Winpak |
CAS | 7732-18-5 |
Sterility | Non-sterile |
Solubility | Completely miscible |
Form | Liquid |
Product Type | Buffer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction