Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
phospho-c-Myc (Ser347) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$182.31 - $463.74
Specifications
Antigen | phospho-c-Myc (Ser347) |
---|---|
Concentration | 0.5 mg/mL |
Applications | Inhibition Assays, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABC146125UL
|
MilliporeSigma
ABC146125UL |
25 μL |
Each for $182.31
|
|
|||||
ABC1461100
|
MilliporeSigma
ABC1461100UL |
100 μL |
Each for $463.74
|
|
|||||
Description
phospho-c-Myc (Ser347) Polyclonal antibody specifically detects phospho-c-Myc (Ser347) in Human samples. It is validated for Peptide Inhibition Assay, Western Blotting.Specifications
phospho-c-Myc (Ser347) | |
Inhibition Assays, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
KLH-conjugated linear peptide corresponding to 10 amino acids surrounding phosphoserine 347 from the C-terminal half of human c-Myc. | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
0.5 mg/mL | |
Polyclonal | |
Purified | |
RUO | |
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title