Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-CHK1 (Ser345) Monoclonal Antibody (S.48.4)
GREENER_CHOICE

Catalog No. PIMA515145
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515145 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515145 Supplier Invitrogen™ Supplier No. MA515145
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

It is not recommended to aliquot this antibody.

CHEK1 (CHK1) is a serine/threonine protein kinase involved in preventing replication when DNA integrity is compromised. CHK1 is a key mediator in the DNA damage-induced checkpoint network. CHK1 is an evolutionarily conserved protein kinase that functions to ensure genomic integrity upon genotoxic stress. When the G2 or S checkpoint is abrogated by the inhibition of CHK1, p53-deficient cancer cells undergo mitotic catastrophe and eventually apoptosis, whereas normal cells are still arrested in the G1 phase. Thus, CHK1 inhibitors can preferentially potentiate the efficacy of DNA damaging agents in cancer cells, and Chk1 is an attractive therapeutic target for cancer treatment, especially since approximately 50% of all human cancers are p53-deficient.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CHK1 (Ser345)
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone S.48.4
Concentration 37 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 50% glycerol, 0.15M NaCl, 100 μg/mL BSA and <0.02% sodium azide; pH 7.5
Gene Chek1
Gene Accession No. O14757, O35280, Q91ZN7
Gene Alias C85740; Cell cycle checkpoint kinase; checkpoint kinase 1; checkpoint kinase 1 homolog; checkpoint kinase-1; Checkpoint, S. pombe, homolog of, 1; CHEK1; CHK1; CHK1 checkpoint homolog; Chk1-S; homolog of Chk1-S; HRD1 Protein; rad27; rad27 homolog; serine/threonine protein kinase CHK1; serine/threonine-protein kinase Chk1; Synoviolin; YBR1742; YBR274W
Gene Symbols Chek1
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding pSer345 of human Chk1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1111, 12649, 140583
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.