Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
phospho DCC (Tyr1418) Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35 - $456.56
Specifications
Antigen | phospho DCC (Tyr1418) |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Dot Blot, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABC1466-25UL
|
MilliporeSigma
ABC146625UL |
25 μL |
Each for $192.35
|
|
|||||
ABC1466-100
|
MilliporeSigma
ABC1466100UL |
100 μL |
Each for $456.56
|
|
|||||
Description
Anti-phospho DCC (Tyr1418) iis a rabit polyclonal antibody that detects Netrin receptor DCC and has been tested for use in Dot Blot, Immunofluorescence, and Western Blotting.Specifications
phospho DCC (Tyr1418) | |
Dot Blot, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis and Cancer | |
NP_036973 | |
A linear peptide corresponding to 15 amino acids surrounding phospho-tyrosine 1418 from the C-terminal region of rat Netrin receptor DCC. | |
Primary | |
Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. |
Please refer to lot specific datasheet. | |
Polyclonal | |
Serum | |
RUO | |
Rat | |
Dcc | |
IgG | |
Unpurified | |
This rabbit polyclonal antibody detects Netrin receptor DCC phosphorylalted on Tyrosine 1418 in rat cells. It targets an epitope within 15 amino acids from the C-terminal region. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title