Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
phospho-E2F1 (Ser375) Rabbit anti-Human, Unconjugated, Clone: 71-10, MilliporeSigma™
Rabbit Monoclonal Antibody
$192.35 - $452.71
Specifications
Antigen | phospho-E2F1 (Ser375) |
---|---|
Clone | 71-10 |
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABE1782-25
|
MilliporeSigma
MABE178225UL |
25 μL |
Each for $192.35
|
|
|||||
MABE1782-100
|
MilliporeSigma
MABE1782100UL |
100 μL |
Each for $452.71
|
|
|||||
Description
Anti-phospho-E2F1 (Ser375), clone 71-10, Cat. No. MABE1782, is a highly specific rabbit monoclonal antibody that targets Transcription factor E2F1 phosphorylated on Serine 375 and has been tested in ELISA and Western Blotting.Specifications
phospho-E2F1 (Ser375) | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Transcription factor E2F1;E2F-1;PBR3;Retinoblastoma-associated protein 1;RBAP-1;Retinoblastoma-binding protein 3;RBBP-3;pRB-binding protein E2F-1 | |
E2F1;RBBP3 | |
IgG | |
Protein G purified | |
Clone 71-10 is a rabbit monoclonal antibody that specifically detects human E2F1 phosphorylated on Serine 375. | |
∽80 kDa observed ( GST fusion protein); 46.92 kDa calculated (without GST). Uncharacterized bands may be observed in some lysate(s). |
71-10 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Epigenetics & Nuclear Function | |
Purified rabbit monoclonal antibody IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
NP_005216.1 | |
KLH-conjugated linear peptide corresponding to 17 amino acids surrounding phosphoserine 375 from the C-terminal region of human transcription factor E2F1. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. | |
Antibodies |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title