Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
phospho-GluR2 (Ser880) Rabbit anti-Rat,Human,Mouse,Monkey,Chimpanzee, KHL, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$189.29 - $481.52
Specifications
Antigen | phospho-GluR2 (Ser880) |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | KLH |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
07294I25UMI
|
MilliporeSigma
07294I25UG |
25 μg |
Each for $189.29
|
|
|||||
07294I100MI
|
MilliporeSigma
07294I100UG |
100 μg |
Each for $481.52
|
|
|||||
Description
phospho-GluR2 (Ser880) Polyclonal antibody specifically detects phospho-GluR2 (Ser880) in Rat,Human,Mouse,Monkey,Chimpanzee samples. It is validated for Western Blotting.Specifications
phospho-GluR2 (Ser880) | |
Polyclonal | |
Purified | |
Neuroscience | |
NP_058957 | |
FRIA2, GLUR2 | |
IgG | |
Affinity Purified | |
Anti-phospho-GluR2 (Ser880) is a rabbit polyclonal antibody that detects glutamate receptor 2 phosphorylated on serine 880. |
Western Blot | |
KLH | |
Rabbit | |
Rat, Human, Mouse, Monkey, Chimpanzee | |
Glutamate receptor 2;Glu-R2;AMPA-selective glutamate receptor 2;GluR-B;GluR-K2;Glutamate receptor ionotropic;AMPA 2;GluA2 | |
KLH-conjugated Linear Peptide corresponding to 9 amino acids surrounding phosphoserine 880 from the the C-terminal region of Rat Glutamate receptor 2. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼110 kDa observed; 98.69 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title