Learn More
Phospho-HICE1 pS70 (Ser70) Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$178.35 - $552.27
Specifications
| Antigen | Phospho-HICE1 pS70 (Ser70) |
|---|---|
| Concentration | 1.22 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2023
![]()
|
Rockland Immunochemicals
600401C52 |
100 μg |
Each for $552.27
|
|
|||||
50-199-2024
![]()
|
Rockland Immunochemicals
600401C52S |
25 μL |
Each for $178.35
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Hice1 pS70 affinity purified antibody is directed against the phosphorylated form of human Hice1 protein at the S70 residue. The antibody is specific for the phosphorylated form of Hice1. Reactivity with non-phosphorylated human Hice1 is minimal by ELISA and western blot. A BLAST analysis was used to suggest cross reactivity with Hice1 from human based on 100% sequence homology with the immunogen. Reactivity against homologues from other sources is not known.
Hice1 contributes to mitotic spindle assembly, maintenance of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.Specifications
| Phospho-HICE1 pS70 (Ser70) | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9BT25 | |
| 93323 | |
| Primary | |
| Antibody |
| 1.22 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| 2410004L22Rik; 4HAUS augmin-like complex, subunit 8; DGT4; HAUS augmin like complex subunit 8; HAUS augmin-like complex subunit 8; HAUS augmin-like complex, subunit 8; Haus8; HEC1/NDC80 interacting, centrosome associated 1; HEC1/NDC80-interacting centrosome-associated protein 1; Hec1-interacting and centrosome-associated 1; HICE1; MGC20533; NY SAR 48; NYSAR48; NY-SAR-48; Sarcoma antigen NY-SAR-48; Sarcoma antigen NY-SAR-48 homolog | |
| HAUS8 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| HAUS8 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.