Learn More
Phospho-IL13RA1 (Tyr405) Polyclonal Antibody, Invitrogen™
Description
Specifications
Specifications
| Antigen | Phospho-IL13RA1 (Tyr405) |
| Applications | Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | IL13RA1 |
| Gene Accession No. | P78552, O09030 |
| Gene Alias | CD213A1, IL-13Ra, NR4, CD213a1 antigen, CT19, IL-13 receptor subunit alpha-1, IL-13R subunit alpha-1, IL-13R-alpha-1, IL-13RA1, IL13 receptor alpha-1 chain, Interleukin 13 receptor |
| Gene Symbols | IL13RA1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.