Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Jak3 (Tyr980, Tyr981) Recombinant Rabbit Monoclonal Antibody (JAK3Y980981-E10), PE, Invitrogen™

Catalog No. p-7215257
Change view
Click to view available options
Quantity:
10 Tests
100 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537449 10 Tests
PIMA537201 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA537449 Supplier Invitrogen™ Supplier No. MA537449
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Janus kinase 3 (JAK3) phosphorylates members of the STAT (signal transduction and activators of transcription) family. JAK3 is predominantly expressed in immune cells and transduces a signal in response to activation of interleukin receptors. In response to a varitety of cytokine or related factors (e.g., interferon, interleukins), JAKs are activated via phosphorylation at 2 adjacent tyrosine residues. The activation of JAKs can lead to the phosphorylation of STAT (signal transducers and activatiors of transcription) proteins, which dimerize and translocate to the nucleus. Once translocated to the nucleus, the STAT proteins can modify transcription of numerous genes, including interferon-stimulated genes. JAK2 is required for the IFN gamma-receptor complex initiation and JAK1 functions as an amplifier. However, active JAK1 may be required for complex responses. Some studies have suggested that the role of JAK2 might be performed by Tyk2 and JAK3, if they were positioned correctly within the IFN gamma-receptor complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Jak3 (Tyr980, Tyr981)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone JAK3Y980981-E10
Conjugate PE
Formulation PBS with 0.20% BSA and 0.09% sodium azide
Gene JAK3
Gene Accession No. P52333
Gene Alias Breast-JAK3; fae; JAK3; JAK-3; JAK3_HUMAN; JAK3B; JAKL; Janus kinase 3; Janus kinase 3 (a protein tyrosine kinase, leukocyte); Janus kinase 3 protein-tyrosine kinase; Janus kinase 3, protein-tyrosine kinase; L JAK; Leukocyte janus kinase; LJAK; L-JAK; RATJAK3; Tyrosine-protein kinase JAK3; wil
Gene Symbols JAK3
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Tyr980/981 of human Jak3.
Purification Method Protein A/G
Quantity 10 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3718
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.