Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-KIF20A (Ser528) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA540224
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA540224 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA540224 Supplier Invitrogen™ Supplier No. PA540224
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-KIF20A (Ser528) Polyclonal Antibody for Western Blot, IHC, ELISA

Mitotic kinesin required for chromosome passenger complex (CPC)-mediated cytokinesis. Following phosphorylation by PLK1, involved in recruitment of PLK1 to the central spindle. Interacts with guanosine triphosphate (GTP)-bound forms of RAB6A and RAB6B. May act as a motor required for the retrograde RAB6 regulated transport of Golgi membranes and associated vesicles along microtubules. Has a microtubule plus end-directed motility. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-KIF20A (Ser528)
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KIF20A
Gene Accession No. O95235
Gene Alias AA415432; GG10_2; KIF20A; kinesin family member 20A; kinesin-like protein 174; Kinesin-like protein KIF20A; mitotic kinesin-like protein 2; MKlp2; RAB6 interacting, kinesin-like (rabkinesin6); rab6-interacting kinesin-like protein; RAB6KIFL; Rabkinesin-6
Gene Symbols KIF20A
Host Species Rabbit
Immunogen Synthesized peptide derived from human KIF20A around the phosphorylation site of S528.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10112
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.