Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-mTOR (Ser2448) Recombinant Rabbit Monoclonal Antibody (mTORS2448-E11), FITC, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
$245.00 - $598.50
Specifications
Antigen | Phospho-mTOR (Ser2448) |
---|---|
Clone | mTORS2448-E11 |
Applications | Flow Cytometry |
Classification | Recombinant Monoclonal |
Conjugate | FITC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA537392
|
Invitrogen™
MA537392 |
10 Tests |
Each for $245.00
|
|
|||||
PIMA537146
|
Invitrogen™
MA537146 |
100 Tests |
Each for $598.50
|
|
|||||
Description
FRAP1 (mTOR) is a serine/threonine kinase that plays a critical role in cellular growth and proliferation. Perturbations in the mTOR/PI3-kinase/AKT pathway are associated with numerous forms of cancer. FRAP1 is also the target of rapamycin and its analogues, which are currently used as immunosuppressants and cancer therapeutics. Mutations affecting the gene results in Smith-Kingsmore syndrome.Specifications
Phospho-mTOR (Ser2448) | |
Flow Cytometry | |
FITC | |
Rabbit | |
Human, Mouse | |
P42345, Q9JLN9 | |
2475, 56717 | |
A synthetic phospho-peptide corresponding to residues surrounding Ser2448 of human phospho mTOR. | |
Primary | |
4°C, store in dark | |
MTOR |
mTORS2448-E11 | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with 0.20% BSA and 0.09% sodium azide | |
2610315D21Rik; AI327068; angiopoietin-like factor CDT6; FK506 binding protein 12-rapamycin associated protein 1; FK506 binding protein 12-rapamycin associated protein 2; FK506-binding protein 12-rapamycin complex-associated protein 1; FKBP12-rapamycin complex-associated protein; FKBP12-rapamycin complex-associated protein 1; FKBP-rapamycin associated protein; FKBP-rapamycin associated protein (FRAP); FKBP-rapamycin-associated protein FRAP; flat; FRAP; FRAP1; FRAP2; Mammalian target of rapamycin; mechanistic target of rapamycin; mechanistic target of rapamycin (serine/threonine kinase); mechanistic target of rapamycin kinase; Mtor; m-TOR; mTORC1; Raft1; Rapamycin and FKBP12 target 1; rapamycin and FKBP12 target-1 protein; rapamycin associated protein FRAP2; rapamycin target protein 1; RAPT1; serine/threonine-protein kinase mTOR; SKS | |
MTOR | |
IgG κ | |
Protein A/G | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title