Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
phospho-NCF1/p47 PHOX (S345) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$191.34 - $481.79
Specifications
Antigen | phospho-NCF1/p47 PHOX (S345) |
---|---|
Concentration | 1.0 mg/mL |
Applications | Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABF180925UG
|
MilliporeSigma
ABF180925UG |
25 μg |
Each for $191.34
|
|
|||||
ABF1809100
|
MilliporeSigma
ABF1809100UG |
100 μg |
Each for $481.79
|
|
|||||
Description
phospho-NCF1/p47 PHOX (S345) Polyclonal antibody specifically detects phospho-NCF1/p47 PHOX (S345) in Human samples. It is validated for Immunofluorescence, Western Blotting.Specifications
phospho-NCF1/p47 PHOX (S345) | |
Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Ovalbumin-conjugated linear peptide corresponding to 16 amino acids from the C-terminal region of human Neutrophil cytosol factor 1 (NCF1) phosphorylated on serine 345. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. |
1.0 mg/mL | |
Polyclonal | |
Purified | |
RUO | |
Purified rabbit polyclonal antibody in buffer containing 10 mM Tris-HCl pH7.5 with 0.01% sodium azide with 10% glycerol. | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title