Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
phospho-p62 (Ser24) Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$182.31 - $461.76
Specifications
Antigen | phospho-p62 (Ser24) |
---|---|
Applications | Dot Blot, ELISA, Peptide Array, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABC204725MI
|
MilliporeSigma
ABC204725UL |
25 μL |
Each for $182.31
|
|
|||||
ABC204710MI
|
MilliporeSigma
ABC2047100UL |
100 μL |
Each for $461.76
|
|
|||||
Description
phospho-p62 (Ser24) Polyclonal antibody specifically detects phospho-p62 (Ser24) in Human,Mouse,Feline,Rhesus Macaque,Rat samples. It is validated for Dot Blot, ELISA, Peptide Array, Western Blotting.Specifications
phospho-p62 (Ser24) | |
Polyclonal | |
Purified | |
Apoptosis and Cancer | |
NP_003891 | |
SQSTM1, ORCA, OSIL | |
IgG | |
Affinity Purified | |
This rabbit polyclonal antibody specifically detects Sequestosome-1 (p62) phosphorylated on serine 24. |
Dot Blot, ELISA, Peptide Array, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Feline, Rhesus Macaque, Rat | |
Sequestosome-1;EBI3-associated protein of 60 kDa;EBIAP;p60;Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa;Ubiquitin-binding protein p62 | |
Linear peptide corresponding to 8 amino acids surrounding phosphoserine 24 in the N-terminal region of human p62. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼62 kDa observed; 47.69 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title