Learn More
Phospho-p90 RSK1 (Ser732) Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$178.35 - $661.74
Specifications
| Antigen | Phospho-p90 RSK1 (Ser732) |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-1998
![]()
|
Rockland Immunochemicals
600401B30 |
100 μg |
Each for $661.74
|
|
|||||
50-199-1999
![]()
|
Rockland Immunochemicals
600401B30S |
25 μL |
Each for $178.35
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for human p90 RSK1 protein phosphorylated at S732. A BLAST analysis was used to suggest cross reactivity with p90 RSK1 from human, rat, mouse, horse, bovine, and dog based on 100% homology with the immunizing sequence. Partially cross reactivity may occur against opossum and chicken based on >90% homology with the immunizing sequence. Cross reactivity with p90 RSK1 pS732 from other sources has not been determined.
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
| Phospho-p90 RSK1 (Ser732) | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Bovine, Canine, Equine, Human, Mouse, Rat | |
| P18653, Q15418, Q63531 | |
| 100057166, 20111, 487353, 533908, 6195, 81771 | |
| Primary | |
| Antibody |
| 1 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| 90 kDa ribosomal protein S6 kinase 1; CLS; dJ590P13.1 (ribosomal protein S6 kinase, 90kD, polypeptide 1); HU-1; HU-2; HU-3; ISPK-1; MAP kinase-activated protein kinase 1; MAP kinase-activated protein kinase 1a; MAPK-activated protein kinase 1; MAPK-activated protein kinase 1a; MAPKAP kinase 1; MAPKAP kinase 1a; MAPKAP kinase 1c; MAPKAPK-1; Mapkapk1a; Mapkapk-1a; MAPKAPK1B; MAPKAPK1C; MAPKAPK-1c; MRX19; OTTHUMP00000004114; OTTMUSP00000010160; p90Rsk; p90-RSK; p90-RSK 1; p90-RSK 2; p90Rsk1; p90-Rsk1; p90RSK2; p90-RSK2; p90-RSK3; p90S6K; pp90RSK1; pp90RSK2; pp90RSK3; Ribosomal protein S6 kinase 2 alpha; ribosomal protein S6 kinase 2 beta; ribosomal protein S6 kinase a, polypeptide 1; ribosomal protein S6 kinase A1; ribosomal protein S6 kinase A1 L homeolog; ribosomal protein S6 kinase alpha 1; Ribosomal protein S6 kinase alpha-1; Ribosomal protein S6 kinase II alpha; ribosomal protein S6 kinase II beta; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase, 90kD, polypeptide 1; Ribosomal protein S6 kinase, 90kD, 1; ribosomal protein S6 kinase, 90kDa, polypeptide 1; ribosomal protein S6 kinase, 90kDa, polypeptide 1 L homeolog; ribosomal protein S6 kinase, polypeptide A1; Ribosomal S6 kinase 1; RP11-393H10.3; RP11-492M19.2; RPS6KA; Rps6ka1; rps6ka1.L; RSK; Rsk1; RSK-1; RSK2; RSK3; RSK-3; S6 kinase II beta; S6 protein kinase (Rsk-1); S6K-alpha; S6K-alpha 1; S6K-alpha-1; S6K-alpha2; S6K-alpha-2; S6K-alpha3; S6KII-alpha; S6KII-beta; wu:fc56h07; XELAEV_18012109mg; zgc:152654 | |
| RPS6KA1 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| RPS6KA1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.