Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-PRAS40 (Thr246) Recombinant Rabbit Monoclonal Antibody (3A1F3)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA558237

Description
Immunogen sequence: LNT(p)SD.
The Akt signaling pathway contributes to the regulation of apoptosis after a variety of cell death signals. AKT1S1, also known as PRAS40, is a proline-rich substrate of the kinase AKT1 and is thought to play a role in neuroprotection mediated by nerve growth factor (NGF) after transient focal cerebral ischemia. AKT1S1 is also a substrate and potential regulator of mammalian target of rapamycin (mTOR), a serine/threonine kinase that regulates cell growth and cell cycle, and a negative regulator of autophagy. Treatment with the insulin-like growth factor-1 (IGF1) can induce the phosphorylation of AKT1S1 via the PI3K/AKT signaling pathway in PC12 cells.
Specifications
Phospho-PRAS40 (Thr246) | |
Recombinant Monoclonal | |
4.5 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q96B36, Q9D1F4 | |
AKT1S1 | |
A synthetic phosphorylated peptide around T246 of human AKT1S1/PRAS40 (NP_001265088.1). | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ELISA, Western Blot | |
3A1F3 | |
Unconjugated | |
AKT1S1 | |
1110012J22Rik; 40 kDa proline-rich AKT substrate; AI227026; AKT1 substrate 1; AKT1 substrate 1 (proline rich); AKT1 substrate 1 (proline-rich); akt1s1; Lobe; Lobel; OTTHUMP00000196756; OTTHUMP00000196760; PKB/Akt substrate 40; Pras; PRAS40; Proline-rich AKT substrate; proline-rich AKT1 substrate 1 | |
Rabbit | |
Protein A | |
RUO | |
67605, 84335 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction