Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho RasGRP2 (Ser116/Ser117) Rabbit anti-Monkey, Mouse, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABS213025UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS2130-25UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS2130-25UG Supplier MilliporeSigma Supplier No. ABS213025UG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-phospho RasGRP2 (Ser116/Ser117), Cat. No. ABS2130, is a rabbit polyclonal antibody that detects RAS guanyl-releasing protein 2 phosphorylated on Ser116/Ser117 and has been tested for use in Immunofluorescence and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho RasGRP2 (Ser116/Ser117)
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine, 10% Ethylene Glycol, pH 7.6 with 0.05% sodium azide.
Gene Alias RAS guanyl-releasing protein 2;Calcium and DAG-regulated guanine nucleotide exchange factor I;CalDAG-GEFI;F25B3.3 kinase-like protein
Gene Symbols Rasgrp2
Host Species Rabbit
Immunogen A linear peptide corresponding to 12 amino acids surrounding phosphorylated Ser116/117 from the N-terminal region of murine RAS guanyl-releasing protein 2.
Molecular Weight of Antigen ∽90 kDa observed; 69.45 kda calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects RAS guanyl-releasing protein 2 phosphorylated on Ser116/117 in Mouse and Monkey cells. It targets an epitope with in 12 amino acids surrounding Ser116/Ser117 from the N-terminal region.
Target Species Monkey, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.