Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Tyrosine Antibody (P-TYR-01), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB500335UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500335UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500335UV Supplier Novus Biologicals Supplier No. NB500335UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Phospho-Tyrosine Monoclonal specifically detects Phospho-Tyrosine in Non-species specific samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Phospho-Tyrosine
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone P-TYR-01
Conjugate DyLight 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Gene Alias 2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, phosphotyrosine, pTyrosine, Tyrosine
Host Species Mouse
Immunogen Phosphotyrosine conjugated to BSA.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The antibody P-Tyr-01 detects tyrosine phosphorylation in activated cells.
Target Species All species
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.