Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Tyrosine Antibody (PY265), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB381839 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB381839 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB381839 Supplier Novus Biologicals Supplier No. NBP308299AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Phospho-Tyrosine Monoclonal antibody specifically detects Phospho-Tyrosine in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Phospho-Tyrosine
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone PY265
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias 2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, phosphotyrosine, pTyrosine, pY20, Tyrosine
Host Species Mouse
Immunogen Phosphotyrosine conjugated to BSA
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience, Phospho Specific, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.