Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
phospho-VE-cadherin (Tyr731) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35 - $510.84
Specifications
Antigen | phospho-VE-cadherin (Tyr731) |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot, Inhibition Assays |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AB1956-I-50UL
|
MilliporeSigma
AB1956I50UL |
50 μL |
Each for $192.35
|
|
|||||
AB1956-I-200
|
MilliporeSigma
AB1956I200UL |
200 μL |
Each for $510.84
|
|
|||||
Description
Anti-phospho-VE-cadherin (Tyr731) is a rabbit polyclonal antibody that detects Cadherin-5 phosphorylated on Tyrosine 731 and has been tested for use in Western Blotting and Peptide Inhibition Assay.Specifications
phospho-VE-cadherin (Tyr731) | |
Western Blot, Inhibition Assays | |
Unconjugated | |
RUO | |
Human | |
CDH5 | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Please refer to lot specific datasheet. | |
Polyclonal | |
Rabbit | |
Cell Structure | |
NP_001786.2 | |
KLH-conjugated linear peptide corresponding to 11 amino acids surrounding phosphorylated tyrosine 731 from human Cadherin-5. | |
Affinity Purified | |
This rabbit polyclonal antibody detects human and murine Cadherin-5 phosphorylated on Tyrosine 731. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title