Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphodiesterase 4A/PDE4A, p Ser686, p Ser688 Antibody, Novus Biologicals™
SDP

Catalog No. NB300635005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300635005 0.05 mg
NB300635025 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300635005 Supplier Novus Biologicals Supplier No. NB3006350.05MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phosphodiesterase 4A/PDE4A Polyclonal specifically detects Phosphodiesterase 4A/PDE4A in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen Phosphodiesterase 4A/PDE4A
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:10000, Immunoprecipitation 1:150
Formulation Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% sodium azide
Gene Accession No. P27815
Gene Alias cAMP-specific 3′-5′-cyclic phosphodiesterase 4A, DPDE2cAMP-specific phosphodiesterase, EC 3.1.4, EC 3.1.4.17, PDE4, PDE46, phosphodiesterase 4A, cAMP-specific, phosphodiesterase 4A, cAMP-specific (dunce, phosphodiesterase 4A, cAMP-specific (dunce (Drosophila)-homologphosphodiesterase E2), phosphodiesterase E2 dunce homolog, Drosophila, phosphodiesterase isozyme 4
Gene Symbols PDE4A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to PKA phosphorylation site on PDE4A long form enzymes. The peptide was synthesized with phosphorylated serine.
Quantity 0.05 mg
Primary or Secondary Primary
Gene ID (Entrez) 5141
Test Specificity This detects all known long-from variants of PDE4A, including PDE4A4, PDE4A5, PDE4A8, PDE4Ax when phosphorylated at the PKA phophorylation site. It will not cross-react with PDE4A phosphorylated at the ERK phosphorylation site. It does not cross-react with PDE4B, PDE4D or any other member of the PDE family.
Target Species Human, Mouse, Rat, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.