Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphodiesterase 4D Rabbit anti-Human, DyLight 650, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB054465 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB054465 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB054465 Supplier Novus Biologicals Supplier No. NBP298613C

Rabbit Polyclonal Antibody

Phosphodiesterase 4D Polyclonal specifically detects Phosphodiesterase 4D in Human samples. It is validated for Western Blot.

Specifications

Antigen Phosphodiesterase 4D
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias cAMP-specific (dunce (Drosophila)-homologphosphodiesterase E3), cAMP-specific 3'-5'-cyclic phosphodiesterase 4D, cAMP-specific phosphodiesterase PDE4D6, DKFZp686M11213, Drosophila), dunce-like phosphodiesterase E3, EC 3.1.4, FLJ97311, HSPDE4D, phosphodiesterase 4D, cAMP-specific, phosphodiesterase 4D, cAMP-specific (phosphodiesterase E3 dunce homolog
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Phosphodiesterase 4D fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5144
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.