Learn More
Description
Specifications
Specifications
| Antigen | Glucose Oxidase |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Dilution | Western Blot 1:500 - 1:3000, ELISA 1:5000 - 1:25000 |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide |
| Gene Alias | Beta D Glucose Oxygen 1 Oxido Reductase, Glucose Oxyhydrase, GOD |
| Host Species | Rabbit |
| Immunogen | Anti-Glucose Oxidase Antibody was produced by repeated immunizations with Aspergillus niger Glucose Oxidase protein. |
| Purification Method | Delipidation and Defibrination |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
