Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoenolpyruvate Carboxylase Antibody, Novus Biologicals™
SDP

Catalog No. NBP244235 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP244235 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP244235 Supplier Novus Biologicals Supplier No. NBP244235
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Phospho Enol Pyruvate Carboxylase in C. elegans, Plant samples, and it is validated for ELISA, Immunohistochemistry, Western Blotting

Phosphoenolpyruvate Carboxylase Polyclonal specifically detects Phosphoenolpyruvate Carboxylase in Plant samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen Phosphoenolpyruvate Carboxylase
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:4000, ELISA 1:10000-1:40000, Immunohistochemistry 1:10-1:500
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. B8XPZ2
Gene Alias PEPC, PEPC 1, PEPC1, PEPCase, PEPCase 1, Phosphoenolpyruvate carboxylase 1, PPC1, PPEPC4
Gene Symbols LOC542372
Host Species Rabbit
Immunogen Phosphoenolpyruvate Carboxylase Antibody was produced by repeated immunizations with maize leaves Phospho-enol-pyruvate Carboxylase. (Uniprot: B8XPZ2)
Molecular Weight of Antigen 109.36 kDa
Purification Method Multi-step
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 542372
Reconstitution Reconstitute with 100 ul deionized water (or equivalent)
Target Species Plant
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.