Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoenolpyruvic acid tris(cyclohexylammonium) salt monohydrate, 97+%, Thermo Scientific™

Catalog No. p-7023596
Click to view available options
Quantity:
0.25 g
1 g

Catalog No. AAA1731701

Supplier: Thermo Scientific Chemicals A1731701

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123334-13-4
Molecular Formula C21H46N3O7P
Molecular Weight (g/mol) 483.587
MDL Number MFCD00004258
InChI Key CWNPPSJOSPTFGQ-UHFFFAOYSA-N
Synonym tri cyclohexylamine salt, 3c6h13n.c3h5o6p.h2o, cyclohexanamine,2-phosphonooxyprop-2-enoic acid,hydrate, phosphoenolpyruvic acid tri cyclohexylamine salt monohydrate, phosphoenolpyruvic acid tris cyclohexylammonium salt hydrate, tris cyclohexylamine phosphoenolpyruvic acid hydrate, 2-phosphonooxy prop-2-enoic acid-cyclohexanamine-water 1/3/1
PubChem CID 71311207
IUPAC Name cyclohexanamine;2-phosphonooxyprop-2-enoic acid;hydrate
SMILES C=C(C(=O)O)OP(=O)(O)O.C1CCC(CC1)N.C1CCC(CC1)N.C1CCC(CC1)N.O

Specifications

Melting Point ∼190°C (decomposition)
Quantity 0.25 g
Formula Weight 483.59 (465.57 Anhydrous)
Chemical Name or Material Phosphoenolpyruvic acid tris(cyclohexylammonium) salt monohydrate
Recommended Storage −30° to −10°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.