Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoglucomutase 5 Antibody (rPGM5/7164), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB155491 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB155491 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB155491 Supplier Novus Biologicals Supplier No. NBP314133AF750
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Phosphoglucomutase 5 Monoclonal antibody specifically detects Phosphoglucomutase 5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Phosphoglucomutase 5
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rPGM5/7164
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias aciculin, EC 5.4.2.2, PGM-RP, PGMRPphosphoglucomutase-like protein 5, phosphoglucomutase 5, Phosphoglucomutase-related protein
Host Species Mouse
Immunogen Recombinant full-length human Phosphoglucomutase 5 protein (Uniprot: Q15124)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5239
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.