Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoglucomutase 5 Rabbit anti-Human, Clone: PGM5/7005R, Novus Biologicals™
SDP

Catalog No. NB177419 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177419 20 μg
NB177418 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB177419 Supplier Novus Biologicals Supplier No. NBP31373120UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Phosphoglucomutase 5 Monoclonal antibody specifically detects Phosphoglucomutase 5 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Phosphoglucomutase 5
Applications Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PGM5/7005R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, ELISA, Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 2-4 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. Q15124
Gene Alias aciculin, EC 5.4.2.2, PGM-RP, PGMRPphosphoglucomutase-like protein 5, phosphoglucomutase 5, Phosphoglucomutase-related protein
Host Species Rabbit
Immunogen Cytoskeletal fraction of the human myometrium tissue extract (Uniprot: Q15124)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5239
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.