Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospholamban, p Ser16 Antibody, Novus Biologicals™
SDP

Catalog No. NB6001186 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001186 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001186 Supplier Novus Biologicals Supplier No. NB6001186
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospholamban (p Ser16) Polyclonal antibody specifically detects Phospholamban (p Ser16) in Human, Bovine samples. It is validated for Western Blot.

Specifications

Antigen Phospholamban (p Ser16)
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml
Formulation 0.1M Tris-Glycine (pH 7.4), 0.15M NaCl and 30% Glycerol with 0.05% Sodium Azide
Gene Accession No. P26678
Gene Alias CMD1PPLBcardiac phospholamban, phospholamban
Gene Symbols PLN
Host Species Rabbit
Immunogen Synthetic peptide corresponding to aa14-25, RA[pS]TIEMPQQAR-C of human cardiac phospholamban with a cysteine added at the C-terminal, conjugated to KLH. Species Sequence Homology: mouse, rat, rabbit, porcine and canine
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5350
Test Specificity Recognizes bovine Phospho-lamban phosphorylated (Ser16) (5-25kD). Additional non-specific bands may be detected at 75 and 100kD in some lysates.
Target Species Bovine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.