Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospholipase B1 Antibody (OTI1D3), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N273386A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273386A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273386A647 Supplier Novus Biologicals Supplier No. NBP273386AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Phospholipase B1 Monoclonal specifically detects Phospholipase B1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Phospholipase B1
Applications Western Blot
Classification Monoclonal
Clone OTI1D3
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ30866, hPLB, phospholipase B1, PLBmembrane-associated
Gene Symbols PLB1
Host Species Mouse
Immunogen Full length recombinant protein of human Phospholipase B1 (NP_694566) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 151056.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.