Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospholipase D2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP223721 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223721 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223721 Supplier Novus Biologicals Supplier No. NBP223721
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospholipase D2 Polyclonal specifically detects Phospholipase D2 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Phospholipase D2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:100, Immunohistochemistry-Paraffin 1:50 - 1:100
Formulation 0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide
Gene Alias Choline phosphatase 2, EC 3.1.4.4, hPLD2, Phosphatidylcholine-hydrolyzing phospholipase D2, phospholipase D2, PLD 2, PLD1C
Gene Symbols PLD2
Host Species Rabbit
Immunogen Within the range amino acids 199-230 of human PLD2 protein were used as the immunogen.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5338
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_b, CRA_c,and CRA_d), 93% homologous in chimpanzee and 81% homologous in rhesus monkey.
Target Species Human, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.