Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospholipid Scramblase 1/PLSCR1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15781620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15781620 20 μL
NBP157816 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15781620 Supplier Novus Biologicals Supplier No. NBP15781620UL

Rabbit Polyclonal Antibody

Phospholipid Scramblase 1/PLSCR1 Polyclonal specifically detects Phospholipid Scramblase 1/PLSCR1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Phospholipid Scramblase 1/PLSCR1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. O15162
Gene Alias Erythrocyte phospholipid scramblase, MmTRA1b, MMTRA1BCa(2+)-dependent phospholipid scramblase 1, phospholipid scramblase 1, PL scramblase 1
Gene Symbols PLSCR1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to PLSCR1(phospholipid scramblase 1) Antibody(against the N terminal of PLSCR1. Peptide sequence MDKQNSQMNASHPETNLPVGYPPQYPPTAFQGPPGYSGYPGPQVSYPPPP.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5359
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.