Learn More
Description
Specifications
Specifications
| Antigen | Phospholipid Scramblase 1/PLSCR1 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.52mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 |
| Formulation | PBS pH7, 1% BSA, 20% Glycerol with 0.01% Thimerosal |
| Gene Alias | Erythrocyte phospholipid scramblase, MmTRA1b, MMTRA1BCa(2+)-dependent phospholipid scramblase 1, phospholipid scramblase 1, PL scramblase 1 |
| Gene Symbols | PLSCR1 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
