Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphomolybdic Acid, Ozone Friendly Readyto Use Spray for TLC, Thermo Scientific™

Catalog No. ac327032400
Click to view available options
Quantity:
240 mL

Catalog No. AC327032400

Supplier: Thermo Scientific Chemicals 327032400

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11104-88-4
Molecular Formula HMoO2P-6
Molecular Weight (g/mol) 159.93
InChI Key FROPCQVXXZDEBU-UHFFFAOYSA-M
Synonym ammonium phosphomolybdate, molybdenum; oxygen 2-; phosphorus 3-; hydroxide, molybdenum hydroxide oxide phosphate, ammoniumphosphomolybdate, molybdophosphoric acid h3pmo12o40 , nickel 2 salt, 12026-57-2 trihydride, molybdophosphoric acid h3pmo12o40 , nickel 2+ salt, molybdophosphoric acid, h3pmo12o40 , cobalt 2 salt, molybdophosphoric acid, h3pmo12o40 , cobalt 2+ salt, 68130-36-9 nickel salt
PubChem CID 159350
IUPAC Name molybdenum;oxygen(2-);phosphorus(3-);hydroxide
SMILES [OH-].[O-2].[P-3].[Mo]

Specifications

Composition TLC spray reagent at 0.1% in isopropanol
Quantity 240 mL
Merck Index 13, 7428
Chemical Name or Material Phosphomolybdic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.