Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphopantothenate-cysteine ligase Antibody (10D11), HRP, Novus Biologicals™
SDP

Catalog No. NB162254 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162254 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB162254 Supplier Novus Biologicals Supplier No. NBP259481H
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Phosphopantothenate-cysteine ligase Monoclonal antibody specifically detects Phosphopantothenate-cysteine ligase in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Phosphopantothenate-cysteine ligase
Applications Western Blot, ELISA
Classification Monoclonal
Clone 10D11
Conjugate HRP
Formulation PBS
Gene Alias FLJ11838, MGC117357, phosphopantothenoylcysteine synthetase, PPC synthetase
Host Species Mouse
Immunogen Phosphopantothenate-cysteine ligase (1-311aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 79717
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.