Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoribosyl Pyrophosphate Amidotransferase Antibody (OTI1B8), Biotin, Novus Biologicals™
SDP

Catalog No. NBP273394B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273394B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273394B Supplier Novus Biologicals Supplier No. NBP273394B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Phosphoribosyl Pyrophosphate Amidotransferase Monoclonal specifically detects Phosphoribosyl Pyrophosphate Amidotransferase in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Phosphoribosyl Pyrophosphate Amidotransferase
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1B8
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ATASE, EC 2.4.2.14, glutamine phosphoribosylpyrophosphatate amidotransferase, Glutamine phosphoribosylpyrophosphate amidotransferase, glutamine PRPP amidotransferase, GPATamidophosphoribosyltransferase, phosphoribosyl pyrophosphate amidotransferase, PRAT
Gene Symbols PPAT
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 42-278 of human Phosphoribosyl Pyrophosphate Amidotransferase (NP_002694) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5471.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.