Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphoric acid, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Clear Glass Bottle
Description
- For Laboratory,Research,or Manufacturing Use
Specifications
Specifications
| Color | White or Colorless |
| Melting Point | 42.35°C |
| Boiling Point | 158°C |
| Density | 1.885g/mL |
| Packaging | Clear Glass Bottle |
| Specific Gravity | 1L = 1.69kg |
| Quantity | 2.5 L |
| Physical Form | Solid or Viscous Liquid |
| Formula Weight | 97.994 |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction