Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphorus(V) bromide, 95%
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Phosphorus(V) bromide, is used in organic chemistry to convert carboxylic acids to acyl bromides
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phosphorus(V) bromide, is used in organic chemistry to convert carboxylic acids to acyl bromides
Solubility
Reacts with water.
Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Phosphorus(V) bromide, is used in organic chemistry to convert carboxylic acids to acyl bromides
Solubility
Reacts with water.
Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Specifications
Specifications
| Boiling Point | 105°C to 107°C (decomposition) |
| Melting Point | ∽105°C (decomposition) |
| Quantity | 100 g |
| UN Number | UN2691 |
| Beilstein | 3903332 |
| Merck Index | 14,7350 |
| Solubility Information | Reacts with water. |
| Formula Weight | 430.5 |
| Percent Purity | 95% |
| Odor | Acrid |
| Show More |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: PHOSPHORUS PENTABROMIDE |
| EINECSNumber | 232-186-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction