Learn More
Phosphorus(V) oxide, ACS, 98% min
Description
Used as a drying and dehydrating agent, condensation reagent in organic synthesis and a laboratory reagent. Used in sugar refining, and in fire extinguishing. Phosphorus pentoxide in DMSO forms Onodera reagent, which oxidizes alcohols. Capable of converting mineral acids to anhydrides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 360°C (sublimation) |
| Melting Point | 340°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 500 g |
| UN Number | UN1807 |
| Merck Index | 14,7355 |
| Synonym | Phosphoric anhydride; Phosphoric oxide |
| Solubility Information | Soluble in H2SO4; Insoluble in acetone and ammonia; Decomposes in water. |
| Formula Weight | 141.94 |
| Show More |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314; Causes severe skin burns and eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: PHOSPHORUS PENTOXIDE |
| EINECSNumber | 215-236-1 |
| RTECSNumber | TH3945000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.