Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoserine Antibody (PSR-45), Novus Biologicals™
SDP

Catalog No. NB600405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600405 Supplier Novus Biologicals Supplier No. NB600405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Phosphoserine Monoclonal specifically detects Phosphoserine in Human, Rat, Rabbit samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Phosphoserine
Applications Western Blot, ELISA
Classification Monoclonal
Clone PSR-45
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, ELISA 1:4000
Gene Alias Phosphoserine; Phosphothreonine
Host Species Mouse
Immunogen Phosphoserine conjugated to Keyhole Limpet Hemocyanin (KLH).
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity By ELISA and dot blot, the antibody reacts specifically with phosphorylated serine, both as free amino acid or conjugated to carriers such as BSA or KLH. No cross-reactivity is observed with non-phosphorylated serine, phosphothreonine, phosphotyrosine, AMP or ATP. This antibody has been used in immunoblotting for the localization of some phosphoserine-containing proteins. Certain proteins known to contain phosphorylated serine may not be recognized by this antibody due to steric hindrance of the recognition site.
Target Species Human, Rat, Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.