Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphothreonine Antibody (18F6), Novus Biologicals™
SDP

Catalog No. NB600878 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600878 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600878 Supplier Novus Biologicals Supplier No. NB600878
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Phosphothreonine Monoclonal specifically detects Phosphothreonine in Non-species specific samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Phosphothreonine
Applications Western Blot, ELISA
Classification Monoclonal
Clone 18F6
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:4000-1:20000
Host Species Mouse
Immunogen Phosphothreonine Antibody (18F6) was produced after repeated immunizations of balb/c mice with phosphothreonine conjugated KLH.
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Signal Transduction
Primary or Secondary Primary
Test Specificity Reactivity is specific for phosphothreonine and minimal cross reactivity is observed against phosphoserine or phosphotyrosine.
Target Species Non-species specific
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.