Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphothreonine Antibody, Novus Biologicals™
SDP

Catalog No. NB11096896 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11096896 0.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11096896 Supplier Novus Biologicals Supplier No. NB11096896
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phosphothreonine Polyclonal specifically detects Phosphothreonine in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Phosphothreonine
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, ELISA 0.5 ug/ml, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 10 ug/mg, Immunohistochemistry-Paraffin
Formulation PBS, 50% Glycerol with 0.09% Sodium Azide
Host Species Rabbit
Immunogen Phosphothreonine conjugated to KLH
Purification Method Affinity Purified
Quantity 0.4 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes proteins phosphorylated on threonine residues. Does not cross-react with phosphotyrosine.
Target Species Non-species specific, Mouse, Non-species specific
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.