Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphothreonine Antibody (RM102) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB301069 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301069 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301069 Supplier Novus Biologicals Supplier No. NBP325980
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Phosphothreonine Monoclonal antibody specifically detects Phosphothreonine in All samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, ChIP assay (ChIP)

Specifications

Antigen Phosphothreonine
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone RM102
Conjugate Unconjugated
Dilution Western Blot : 1:500 -1:2000, Flow Cytometry : 1:10 - 1:1000, ELISA, Immunohistochemistry : 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence : 1:100 - 1:500, Immunoprecipitation : 1:100-1:500, Chromatin Immunoprecipitation (ChIP) : 1:100-1:500
Formulation PBS
Gene Alias C4H10NO6P,O-phospho-L-threonine,Phospho-threonine,pThr,Threonine
Host Species Rabbit
Immunogen Mixture of Phosphothreonine-BSA conjugate and a Phosphothreonine containing peptide
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species All species
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.